Suction Samplers, Mini ViscoSampler, Bürkle
Fournisseur: BURKLE INC
Certificats
A propos de cet article
These suction samplers are ideal for viscous substances such as pastes, mud, creams, oils.
Suction Samplers, Mini ViscoSampler for viscous substances such as pastes, mud, creams, oils. The sample is drawn in using the pump rod and directly conveyed to the sampling container. Easy cleaning for flawless sampling. Depending on the viscosity, the removable suction tip can be fitted with suction drills of 2 mm to 6 mm.
- Easy to clean
- Ideal for pharmaceutical and cosmetics industry
- Sterilizable
- Stainless steel V4A (1.4404)/ AISI 316L
The sample is drawn in using the pump rod and directly conveyed to the sampling container. Easy cleaning for flawless sampling. Depending on the viscosity, the removable suction tip can be fitted with suction drills of 2 mm to 6 mm.
Customers Who Bought This Also Bought