Stop-It Metal Tubing Clamps, Bürkle
Supplier: BURKLE INC
Certificates
About this item
Tubing clamps made of galvanised steel and black anodised aluminium.
- Stepless flow regulation
- Heat resistant
- For hose diameters up to max. 30 mm
Used to regulate or stop the flow in tubing with liquids or gases.
Customers Who Bought This Also Bought