LaboPlast Funnel Stands for Two or Four Funnels, Bürkle
Fournisseur: BURKLE INC
Certificats
A propos de cet article
Sturdy and stable funnel stand, resistant to chemicals, shatter-proof and with continuously variable height adjustment.
- For funnels Ø 40 - 180 mm (1.57 to 7.09")
- Additional funnel holders are available as accessories
The funnel holders can be used with reducing inserts for small funnels. The funnel stand also makes an excellent filtering stand. Innovative magnet concept, no clamping screws are necessary. The height of the funnel holder can be adjusted easily and flexibly with one hand.
Packaging: One funnel stand per package.
Customers Who Bought This Also Bought