A propos de cet article
Solvent pump foot operated for filling of high purity liquids such as solvents. All medium-contacting parts are made of stainless steel (1.4301) and PTFE. Due to the barrel screw joint and the stopcock, the container and the pump can be hermetically sealed. Unwanted vapours can therefore not escape during the filling process. Massive, long-life industrial version. Clip connection for fast connection and disconnection of the air hose.
Solvent pump foot operated for filling of high purity liquids such as solvents. All medium-contacting parts are made of stainless steel (1.4301) and PTFE. Due to the barrel screw joint and the stopcock, the container and the pump can be hermetically sealed. Unwanted vapours can therefore not escape during the filling process. Massive, long-life industrial version. Clip connection for fast connection and disconnection of the air hose.
- Immersion tube can be steplessly adjusted to the barrel depth
- For barrels up to 220 litres
- Suitable for steel barrels with R 2” thread
Spécifications
- Catalog No:
- 10217-240
- 10217-244
- Flow rate:
- 30 L/min
- 30 L/min