Tubing Clamps, PVDF, Bürkle
Supplier: BURKLE INC
Certificates
About this item
The micro adjustment screw enables the flow of liquids to be controlled delicately and smoothly or stopped completely.
- PVDF with galvanised steel hook
- Heat resistant up to +160 °C
- Various sizes available for tubing from 1 - 20 mm external diameter
- Available in four colours
The clamp can also be mounted on existing tube connections. The clamp's closure can be opened in any position, without screwing on and off.
Customers Who Bought This Also Bought