WHEATON® Arsine Generator Apparatus, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Arsine Generator is used for arsenic analysis using silver diethyldithiocarbamate (SDDC) colorimetric method
The design eliminates the ball and socket joint connecting the scrubber and absorber components. Assembled apparatus is freestanding and easy to use. It is manufactured from Wheaton 33 low extractable borosilicate glass that conforms
Customers Who Bought This Also Bought