To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Adhesion G Protein-Coupled Receptor E1
- Clonality:Monoclonal
- Clone:BM8.1
- Gene ID:13733
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Antigen symbol:ADGRE1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- Size:100 µg
- Antigen synonyms:Ly71|Gpf480|F4/80|TM7LN3|DD7A5-7|EGF-TM7|Emr1
- Storage temperature:2-8°C
- Tested applications:Flow Cytometry
- Cat. no.:10208-742
- Supplier No.:70-4801-U100
Spécifications
A propos de cet article
The BM8.1 antibody is specific for mouse F4/80 antigen, a 125 kDa transmembrane protein widely expressed by members of the mononuclear phagocyte system and considered to be a key marker for mature macrophage cells. F4/80 is differentially expressed during myeloid cell development, and may be regulated by certain cytokines within the tissue microenvironment. Other cell types shown to express this antigen include Langerhans cells, Kupffer cells and dendritic cell subsets. BM8.1 is widely used together with antibodies to CD115 (c-fms), CD11b and CD11c to identify myeloid / macrophage cells by flow cytometry.
Type: Primary
Antigen: ADGRE1
Clonality: Monoclonal
Clone: BM8.1
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse