Coliwasa Trumpet, Disposable, Wheaton®
# de catalogue 66174-235
Fournisseur: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Spécifications
- Description:Coliwasa Trumpet
- Cat. no.:66174-235
- Supplier No.:885235
Spécifications
A propos de cet article
Use for sampling sludge or semi-solid material.
- Disposable glass
Disposable glass coliwasa will bore through semi-solid sludge type samples. Liquid and sludge material will remain in the coliwasa due to its tight PTFE resin seal. Tube is 101.6 cm long with 200 ml capacity.
Customers Who Bought This Also Bought