OTAL Foot Pumps, Bürkle
Fournisseur: BURKLE INC
Certificats
A propos de cet article
Delivery tubes made of PP are resistant against weak acids, caustic solutions and detergents.
Delivery tubes made of PVDF are especially for acids and caustic solutions; not for strongly oxidizing acids and caustic solutions.
Delivery tubes made of stainless steel 304 are for solvents and liquids of VbF Hazard Class AIII (not easily flammable).
Battery pump for safe filling of car batteries with acid or distilled water.
- Simple handling
- Very good pump capacity due to large volume foot bellows
- With universal bungs made of soft PVC, for container openings of 30 - 70 mm Ø
Spécifications
- Catalog No:
- 10216-942
- 10216-944
- 10216-946
- 10216-948
- 10216-950
- 76501-446
- 10216-938
- 10216-940
- Flow rate:
- 30 L/min. (1.05 cfm)
- 12 L/min. (0.4 cfm)
- 20 L/min. (0.7 cfm)
- 26 L/min. (0.9 cfm)
- 5 L/min. (0.1 cfm)
- 14 L/min. (3.70 gpm)
- 20 L/min. (0.7 cfm)
- 26 L/min. (0.9 cfm)