Kuderna-Danish Evaporative Concentrators with Clear-Seal™ Joints, Wheaton
# de catalogue 74830-064
Fournisseur: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Spécifications
- Description:Kuderna-Danish Concenterator, Complete (270 mm Synder Column, Flask, Receiving Vessel and Two Clear-Seal™ Joints)
- Cat. no.:74830-064
- Capacity:500 ml
- Supplier No.:297883
Spécifications
A propos de cet article
This unit is designed for the concentration of trace amounts of analytes in an organic solvent prior to analysis
It consists of a 270 mm three-ball Snyder column, a 500 ml flask, and a 10 ml receiving vessel.
Customers Who Bought This Also Bought