LaboPlast® Laboratory Trays, Bürkle
Supplier: BURKLE INC
Certificates
About this item
Trays are suitable as a spill trough for environmentally dangerous, aggressive substances, for experiments, as an etching tray, as an instrument holder, a medicine tray, a photographic development basin, a goods presentation tray and a display tray for foodstuffs.
- Trays can be nested
- Temperature resistent up to 60 °C (briefly up to 80 °C)
Tray are not suitable for temperature controlled cupboards or dishwashers.
Customers Who Bought This Also Bought