LaboPlast® Spray Bottles with Pump Vaporizer, Bürkle
Fournisseur: BURKLE INC
Certificats
A propos de cet article
Spray bottle are made of HDPE, spray pump made of PP and stainless steel.
Liquids can be finely vapourised at the push of a button in an environmentally compatible way. The transparent bottles make identification of contents easy. The protective cover prevents unintentional pressing of the pump and acts as a dust cover.
Customers Who Bought This Also Bought