WHEATON® Arsine Generator Apparatus with Standard Taper 24/40 Clear-Seal™ Joint, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Arsine Generator with standard taper 24/40 Clear-Seal™ Joint engineered for arsine analysis.
- Arsine analysis done using the silver diethyldithiocarbamate (SDDC) colorimetric method
- The arsine absorber has a socket joint
- Made from WHEATON 33 low extractable borosilicate glass
The scrubber tube has a standard taper 24/40 Clear-Seal™ joint and a spherical 12/2 ball joint held together by a size 12 stainless steel ball joint clamp.
Certifications: Meets ASTM/USP and EPA specifications. WHEATON 33 low extractable borosilicate glass conforms to ASTM E 438 Type I, Class A requirements.
Customers Who Bought This Also Bought