ContactUsLinkComponent
Corning® 384-Well Clear Flat Bottom Fibronectin Coated Microplates, Corning
Corning® 384-Well Clear Flat Bottom Fibronectin Coated Microplates, Corning
# de catalogue 89235-028
Fournisseur:  Corning
Product available on GSA Advantage®
PLATE 384W FLAT FIBER CLR CS100
Corning® 384-Well Clear Flat Bottom Fibronectin Coated Microplates, Corning
# de catalogue 89235-028
Fournisseur:  Corning
Product available on GSA Advantage®
Spécifications
  • Description:
    384-well plate, clear, fibronectin coated, with lid
  • Cat. no.:
    89235-028
  • Supplier No.:
    3847
Spécifications
A propos de cet article
PS, fibronectin coated, with lid. Flat bottomed plates suitable for high throughput applications.
Customers Who Bought This Also Bought