Celstir® Suspension Culture Flasks, Wheaton
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Four arm flask with fluoropolymer resin lined sidearm caps.
- Even stirring action
- Each sidearm measures 45 mm
- Autoclavable
Customers Who Bought This Also Bought