Micro-Tissue Grinder Kit, Wheaton
Catalog # 62400-800
Supplier: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Specifications
- Description:Complete kit
- Cat. no.:62400-800
- Supplier No.:358204
Specifications
About this item
Micro tissue grinder kit is ideal for sterile homogenization processes.
- Autoclavable
- Replacement parts available separately
- Made from 33 low extractable borosilcate glass
The micro tissue grinder with screw cap can be used for tissue grinding as well as for additional procedures.
Delivery: It is supplied with 7 micro tissue grinders and the kit is packed in a high density polyethylene case with foam inserts.
Customers Who Bought This Also Bought