ContactUsLinkComponent
Micro Tissue Grinder with PTFE Spacer, Wheaton
Micro Tissue Grinder with PTFE Spacer, Wheaton
# de catalogue 62400-890
Small Business Enterprise
Micro Tissue Grinder Kit, 0.2mL Tissue Grinder
Micro Tissue Grinder with PTFE Spacer, Wheaton
# de catalogue 62400-890
Small Business Enterprise
Spécifications
  • Capacity:
    0.2 ml
  • Ø:
    7 mm
  • Description:
    Grinding Chamber and Smooth Pestle with PTFE Spacer, length 33 mm
  • Cat. no.:
    62400-890
  • Overall Length:
    115 mm
  • Supplier No.:
    357848
Spécifications
A propos de cet article
PTFE spacer aligns the pestle and reduces aerosoling of the product.
  • Includes a finely ground glass pestle and tube
  • Made from low extractable borosilicate glass
  • Autoclavable
Customers Who Bought This Also Bought