Gaumard® Gynecologic Simulator
Fournisseur: American 3B Scientific
Certificats
A propos de cet article
Detailed Gynecologic Trainer
- Full size female lower torso with relevant internal anatomic landmarks
- Minilaparotomy
- Interchangeable cervixes and uteri
This gynecologic simulator represents a female adult lower body with relevant internal anatomical landmarks and anatomically accurate female genital organs, fimbriae and ovaries. Various pelvic examinations possible with this gynecologic simulator include laparoscopy, mini-laparotomy as well as recognition of pathologies and placing of contraceptive devices can be trained.
Customers Who Bought This Also Bought