WHEATON® Crimpers and Decappers, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
These crimpers are used for flip cap seals and for aluminum seals sizes 16.5, 28.5, and 30 mm.
- 1 year warranty
- Not autoclavable
Customers Who Bought This Also Bought