Magna Flex™ Microcarrier Spinner Flask, Wheaton
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Designed for microcarrier cultures with a magnetic stirrer
- Gentle stirring action provided by a bulb shaped glass impeller
- Flasks provides head space ratio of 1:1 for ideal gas exchange
Made from Autoclavable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
Customers Who Bought This Also Bought