Celstir® Suspension Culture Flasks with Jacketed Double Sidearm, Wheaton
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Designed for suspension cell culture with a magnetic stirrer.
- Inlet and outlet hose barb connections for 0.25” (6.35 mm) ID tubing
- 360° water jacket for precise temperature control
- Autoclavable borosilicate glass
Certifications: Conforms to USP Type I and ASTM E 438 Type 1, Class A requirements
Customers Who Bought This Also Bought