ContactUsLinkComponent
Gaumard® Male And Female Catheterization And Ostomy Care Simulator
Gaumard® Male And Female Catheterization And Ostomy Care Simulator
# de catalogue 470119-386
Fournisseur:  American 3B Scientific
SIMULATOR ADV PATIENT CARE M AND FM
Gaumard® Male And Female Catheterization And Ostomy Care Simulator
# de catalogue 470119-386
Fournisseur:  American 3B Scientific
Spécifications
  • Description:
    Catheterization And Ostomy Care Simulator
  • Cat. no.:
    470119-386
  • Supplier No.:
    W45068
Spécifications
A propos de cet article
2 Simulators In 1
  • Complete catheterization training
  • Interchangeable, realistic male organ
  • Fully functional stomas connected to removable, replaceable internal tanks
This “two-in-one” simulator allows:
Catheterization: The interchangeable male and female genital organs feature a modular urethral valve to prevent fluid leakage.
Ostomy care: 3 stomas (transverse colostomy, ileostomy, suprapubic) may be dilated and allow practice of irrigation procedures as well as application of ostomy bags. Each stoma is connected to an internal tank.
Supplied with carrying bag.
Customers Who Bought This Also Bought