Laerdal® Baby Hippy Trainer
# de catalogue 470119-308
Fournisseur: American 3B Scientific
Spécifications
- Description:Baby Hippy
- Cat. no.:470119-308
- Supplier No.:W47060
Spécifications
A propos de cet article
Neonatal Skills
- Practice hip setting and assessment procedures
- Natural feel simulator
Representation of the lower torso and limbs of a newborn female with: Dislocated left hip for practice of the Ortolani Jerk-Sign and Lax right hip for performing the Barlow maneuver.
Customers Who Bought This Also Bought