Gaumard® Gynecologic Simulator
# de catalogue 470119-280
Fournisseur: American 3B Scientific
Spécifications
- Description:Gynecologic Simulator
- Cat. no.:470119-280
- Supplier No.:W45024
Spécifications
A propos de cet article
Full Purpose Gynecologic Trainer
- Full size female adult lower body with removable soft outer skin
- Uterus and cervix feature patented “screw” design for fast and easy change-out
- Includes soft carrying bag
The full size female adult lower body manikin simulator gives students and educators a graphic experience and training with gynecologic exercises such as vaginal speculum examination, bi-manual pelvic examination, IUD insertion techniques, diaphragm sizing and fitting, uterine sounding, and viewing of normal and abnormal cervices.
Customers Who Bought This Also Bought