Gaumard® Newborn Patient Care Manikin
Fournisseur: American 3B Scientific
Certificats
A propos de cet article
Advanced Infant Nursing Care Manikin
- Heel stick and finger prick technique
- Soft upper body skin over torso for “baby like” feel
- Bathing and bandaging activity
- Intramuscular injection in upper thigh
- Interchangeable genitalia
- Male and female catheterization
- Enema administration
This manikin simulates a 0-8 week old newborn with rotatable soft arms and legs. It allows training of basic and advanced pediatric patient care procedures such as: Bathing and bandaging, Tube feeding and gastric suction (movable tongue), Heel stick and finger prick practice, Male and female catheterization, Enema administration. Supplied with T-Shirt, diaper and carrying bag.
Customers Who Bought This Also Bought