3B Scientific® Skeleton Chart
# de catalogue 470120-010
Fournisseur: American 3B Scientific
Spécifications
- Length:50 cm
- Width:67 cm
- Description:Skeleton Chart
- Cat. no.:470120-010
- Finish:Unlaminated
- Supplier No.:VR1113UU
Spécifications
A propos de cet article
This colorful anatomical poster details all the parts of the human skeleton.
- UV Resistant Paper Resists Fading
- Available With Double Sided Lamination To Prevent Curling
The anatomy of the skeleton is labeled on a realistic picture and makes for an easy study of the bones of the body. This human skeleton poster is a great addition to any classroom or doctor's office and is a useful tool for patient education.
Caution: UV Resistant Paper Resists Fading
Available With Double Sided Lamination To Prevent Curling
Customers Who Bought This Also Bought