3B Scientific® Nervous System Chart
# de catalogue 470120-192
Fournisseur: American 3B Scientific
Spécifications
- Length:50 cm
- Width:67 cm
- Description:Nervous System Chart
- Cat. no.:470120-192
- Finish:Unlaminated
- Supplier No.:VR1620UU
Spécifications
A propos de cet article
This anatomical poster of the human nervous system comes in full colorful detail.
- Convenient poster size 50 x 67 cm (20 x 26'')
- Anatomy of the human Nervous System is printed on premium glossy (200g) paper
The poster details the entire human nervous system including the human brain and spinal cord.
Customers Who Bought This Also Bought