3B Scientific® Brain Ventricle
# de catalogue 470119-062
Fournisseur: American 3B Scientific
Spécifications
- Description:Brain Ventricle
- Height:14 cm
- No. of Keyed Items:17
- Cat. no.:470119-062
- Length:14 cm
- Width:11 cm
- Supplier No.:VH410
- No. of Parts:1
Spécifications
A propos de cet article
This model shows both side ventricles of the brain, the third and fourth ventricle and the Aquaeductus cerebri (Sylvius).
- 5X ife size
- 3 year manufacturers warranty
- Includes 3B smart anatomy
The brain ventricles are life-size. This brain ventricle model is delivered on stand.
BodyTextAccessories: For information about 3B Smart Anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought