3B Scientific® Liver With Gall Bladder, Pancreas And Duodenum
# de catalogue 470119-164
Fournisseur: American 3B Scientific
Spécifications
- Description:Liver With Gall Bladder, Pancreas And Duodenum
- Height:5 cm
- No. of Keyed Items:35
- Cat. no.:470119-164
- Length:21 cm
- Width:19 cm
- Supplier No.:VE315
- No. of Parts:1
Spécifications
A propos de cet article
Demonstrating the anatomy of the liver has never been easier.
- Unit Illustrates the Liver, Gall Bladder, Pancreas, and Duodenum's Anatomy
- Three-Year Manufacturer's Warranty
- Model Delivered on Baseboard
- Includes 3B Smart Anatomy
The unit illustrates the anatomy of the liver with the gall bladder, pancreas, and duodenum.
The model is delivered on baseboard.
BodyTextAccessories: For information about 3B Smart Anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought