3B Scientific® Heart On Diaphragm
# de catalogue 470119-134
Fournisseur: American 3B Scientific
Spécifications
- Description:Model Heart On Diaphragm 10Pt
- Height:41 cm
- No. of Keyed Items:44
- Cat. no.:470119-134
- Length:28 cm
- Width:34 cm
- Supplier No.:VD251
- No. of Parts:10
Spécifications
A propos de cet article
This detailed heart depicts the structures of the diaphragm.
- Three-Year Manufacturer's Warranty
- Removable Parts Include the Esophagus, Trachea, Superior Vena Cava, and More
- Heart Depicts Structures of the Diaphragm
- Includes 3B Smart Anatomy
The following parts of the heart can be removed: esophagus, trachea, superior vena cava, aorta, pulmonary artery stem, both atrium walls, and both ventricle walls.
The diaphragm includes a three-year manufacturer's warranty.
BodyTextAccessories: For information about 3B Smart Anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought