3B Scientific® Tongue And Mouth Floor
# de catalogue 470119-096
Fournisseur: American 3B Scientific
Spécifications
- Description:Tongue And Mouth
- Height:17 cm
- Cat. no.:470119-096
- Length:24 cm
- Width:18 cm
- Supplier No.:T12010
- No. of Parts:4
Spécifications
A propos de cet article
Large Size for Easy Examination.
- Three-Year Manufacturer's Warranty
- Removable Base
This tongue model shows the lower jaw up to the second molar and the tongue with mouth-floor musculature in median section and the right sublingual and submandibular gland.
Customers Who Bought This Also Bought