3B Scientific® Femoral Fracture And Hip Osteoarthritis
# de catalogue 470118-992
Fournisseur: American 3B Scientific
Spécifications
- Description:Femoral Fracture And Hip Osteoarthritis
- Cat. no.:470118-992
- Supplier No.:A88
- No. of Parts:1
Spécifications
A propos de cet article
This hip joint model shows the right hip joint of an elderly person in half natural size.
- Illustrate Movement At The Shoulder Joint
- Removable Muscles
- 3 Year Manufacturers Warranty
- Includes 3B Smart Anatomy
In addition to the external anatomy of the hip joint, a frontal section through the femoral neck is shown in relief on the base. The hip joint model shows the femoral fractures that occur most commonly in practice as well as typical wear and tear symptoms of the hip joint (coxarthrosis or hip osteoarthritis).The following fractures are shown on the hip joint: Medial femoral neck fracture, Lateral femoral neck fracture, Fracture through the trochanteric region (pertrochanteric femoral fracture, Femoral shaft fracture, Fracture of the greater trochanter and much more.
BodyTextAccessories: For information about 3B Smart Anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought