3B Scientific® Diseases Of The Esophagus
# de catalogue 470119-156
Fournisseur: American 3B Scientific
Spécifications
- Description:Diseases Of The Esophagus
- Height:19 cm
- No. of Keyed Items:6
- Cat. no.:470119-156
- Length:14 cm
- Width:10 cm
- Supplier No.:K18
- No. of Parts:1
Spécifications
A propos de cet article
This vivid esophagus model designed to educate patients and students is a life-size frontal section that reaches from the lower part of the esophagus to the upper part of the stomach.
- Life Size Depiction
- 2 Available Options
- 3 Year Manufacturers Warranty
- Includes 3B Smart Anatomy
The following illnesses are replicated in the diseases of the esophagus model: Reflux esophagitis, Ulcer, Barrett's Ulcer, Esophageal carcinoma, Esophageal varices and Hiatal hernia. This high quality diseases of the esophagus anatomy model is mounted on base.
BodyTextAccessories: For information about 3B Smart Anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought