Laerdal® Infant I.V. Training Arm
Catalog # 470120-402
Supplier: American 3B Scientific
Specifications
- Description:Infant IV Training Arm
- Cat. no.:470120-402
- Supplier No.:W19564
Specifications
About this item
Use Alone, Or As Part Of The Nursing Baby Sim
- Venipuncture possible in antecubital fossa and dorsum of hand
- Simulated blood may be infused for realistic flashback
Lifelike reproduction of an infant arm for mastering extremity venipuncture procedures and fluid administration in the superficial veins of the hand. Phlebotomy and Venipuncture are possible in antecubital fossa and dorsum of the hand. Fluid may be infused for realistic flashback.
Customers Who Bought This Also Bought