Laerdal® Baby 'Stap' Trainer
# de catalogue 470120-398
Fournisseur: American 3B Scientific
Spécifications
- Description:Baby Stap
- Cat. no.:470120-398
- Supplier No.:W19562
Spécifications
A propos de cet article
Positioned For The Practice Of Lumbar Puncture Techniques
- Lateral decubitus position
- Upright position
- Realistic interchangeable spine with spinal cord may be palpated for location of correct puncture site
- Fluid may be infused
Lateral decubitus position. Realistic interchangeable spine with spinal cord may be palpated for location of correct puncture site. Fluid may be infused.
Customers Who Bought This Also Bought