3B Scientific® Inguinal Hernia
# de catalogue 470119-176
Fournisseur: American 3B Scientific
Spécifications
- Description:Inguinal Hernia
- Height:19 cm
- No. of Keyed Items:26
- Cat. no.:470119-176
- Length:14 cm
- Width:10 cm
- Supplier No.:H13
- No. of Parts:1
Spécifications
A propos de cet article
This natural-sized, graphic model shows the anatomical structures of a male groin with an indirect inguinal hernia, opened in layers.
- 4 progressively more detailed models
- 3 year manufacturers warranty
- Includes 3B smart anatomy
Two diagrammatic illustrations on the base allow for a comparison of direct and indirect hernia. A useful model to help inform patients before undergoing surgical interventions. Mounted on base.
BodyTextAccessories: For information about 3B smart anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought