Heart And Breath Sounds Simulator
Fournisseur: American 3B Scientific
Certificats
A propos de cet article
This hand-held device simulates both heart and breath sounds and allows the determination of these by plugging in one of seven different menus, each containing 16 different sounds.
- Multiple Cartridges Make It Easy to Change Sounds
- All Students Hear the Same Sounds for Ease of Identification
- Flexible and Adaptable Training
The Heart and Breath Sounds Simulator unit includes two external sounders with which students can auscultate the sounds with a stethoscope. Linking additional stethoscope sounders, up to 15 students can auscultate at the same time. A loud speaker can also be connected.
Additionally, the unit has a volume control and two LEDs that illuminate during systole (heart menu) and expiration (breath menu). Virtually maintenance-free, the unit is powered by a 9V battery.
Customers Who Bought This Also Bought