3B Scientific® MICROanatomy™ Digestive Tract
# de catalogue 470119-150
Fournisseur: American 3B Scientific
Spécifications
- Description:Digestive Tract
- Height:19 cm
- No. of Keyed Items:42
- Cat. no.:470119-150
- Length:30 cm
- Width:26 cm
- Supplier No.:K23
- No. of Parts:1
Spécifications
A propos de cet article
Histologic view of four digestive regions.
- Three-Year Manufacturer's Warranty
- Model Displays the Structure of the Esophagus, Stomach, and Small and Large Intestine's Fine Tissues
- Front and Back Sides of the Model Display Different Elements of the Digestive System
- Includes 3B Smart Anatomy
The front of the MICROanatomy Digestive System model, from top to bottom, shows a magnified view in the histological section of the individual sections of the digestive system and their fine-tissue structures.
On the back of the digestive system model, highly magnified views of didactically interesting areas of each of the digestive system sections shown on the front are emphasized.
BodyTextAccessories: For information about 3B Smart Anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought