3B Scientific® Skin Cancer Model
# de catalogue 470119-038
Fournisseur: American 3B Scientific
Spécifications
- Description:Skin Cancer
- Height:12 cm
- No. of Keyed Items:26
- Cat. no.:470119-038
- Length:14 cm
- Width:10 cm
- Supplier No.:J15
- No. of Parts:1
Spécifications
A propos de cet article
This 3B Scientific® Skin Pathology model shows healthy skin and 5 different stages of malignant melanoma on the front and back, enlarged 8 times. In the top view of the skin cancer model, the individual stages of externally visible skin changes are shown, allowing for an assessment according to the 'ABCDE' criteria.
- Magnetic assembly
- Shows micro-anatomical structure from different parts of the body
- 3 year manufacturers warranty
- Includes 3B smart anatomy
The sides of the skin cancer model show the various levels of invasion into the skin layers according to Clark (I-V) and the tumor thickness according to Breslow (in mm). 5 original color illustrations on the base of the skin cancer model show various types of malignant melanomas. The skin cancer model comes mounted on a base.
BodyTextAccessories: For information about 3B smart anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought