3B Scientific® Female Pelvis And Femur Heads
# de catalogue 470119-180
Fournisseur: American 3B Scientific
Spécifications
- Description:Female Pelvis And Femur Heads
- Height:18 cm
- Cat. no.:470119-180
- Length:27 cm
- Width:27 cm
- Supplier No.:A62
- No. of Parts:3
Spécifications
A propos de cet article
This realistic pelvic skeleton model consists of hip bone, sacrum with coccyx and 2 lumbar vertebrae as well as movable symphysis.
- Supplied on a flexible stand
- Three years manufacturers warranty
- Includes 3B smart anatomy
BodyTextAccessories: For information about 3B smart anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought