3B Scientific® Classic Heart With Thymus
# de catalogue 470119-122
Fournisseur: American 3B Scientific
Spécifications
- Description:Heart With Thymus
- Height:20 cm
- No. of Keyed Items:41
- Cat. no.:470119-122
- Length:13 cm
- Width:13 cm
- Supplier No.:G08/1
- No. of Parts:3
Spécifications
A propos de cet article
Highly detailed 2-part heart at a price you will love. The front heart wall is detachable to reveal the chambers and valves inside. Heart just slightly smaller than life-size with exquisite anatomical detail throughout, including thymus.
- Clear heart highlights conducting system
- Removable base
- Three years manufacturers warranty
- Includes 3B smart anatomy
Great model of the anatomy of the human heart. Stand included with heart.
BodyTextAccessories: For information about 3B smart anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought