Spécifications
- Description:Combination Skull
- Height:19 cm
- Cat. no.:470118-902
- Length:15 cm
- Width:13 cm
- Supplier No.:A282
- No. of Parts:8
Spécifications
A propos de cet article
The right, transparent skull half allows the study of important anatomical details, such as the location of the paranasal sinuses. Therefore, in combination with the left, bony skull half, a direct transfer of the otherwise invisible structures becomes easy and uncomplicated.
- Clear and opaque halves showing differing structures
- Includes brain
- Three years manufacturers warranty
- Includes 3B smart anatomy
The transparency of the jaw in this skull allows an exceptional view onto the periodontal pockets and roots. The teeth from the skull are removable for detailed studies. In addition, the external masticator muscles (masseter and temporal muscles) are represented on the bony skull half.
To demonstrate masticator movement, the lower jaw in the skull is mounted flexibly. The skull can be disassembled into both halves of the skullcap and base of skull, the nasal septum, the complete mandible and both masticator muscles.