3B Scientific® Eye In Bony Orbit
# de catalogue 470119-100
Fournisseur: American 3B Scientific
Spécifications
- Description:Eye In Bony Orbit
- Height:21 cm
- No. of Keyed Items:23
- Cat. no.:470119-100
- Length:19 cm
- Width:19 cm
- Supplier No.:F11
- No. of Parts:7
Spécifications
A propos de cet article
Removable parts of this anatomical human eye model include: Upper half of the sclera with cornea and eye muscle attachments, both halves of choroid with iris and retina, lens and vitreous humour.
- 5× life size
- Includes base
- Three years manufacturers warranty
- Includes 3B smart anatomy
This eye model is great for studying the anatomy of the human eye! Eye on base of bony orbit.
BodyTextAccessories: For information about 3B smart anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought