3B Scientific® Dentition Development Set
# de catalogue 470119-066
Fournisseur: American 3B Scientific
Spécifications
- Description:Dentition Development Set
- Cat. no.:470119-066
- Supplier No.:D20
Spécifications
A propos de cet article
The dentition development model is cast from a natural specimen, 4 upper and lower jaw halves, 4 different stages of dentition development: New born, Approx. 5-year old child, Approx. 9-year old child, Young adult Dentition development model on a stand.
- 3× life size
- Includes sectioned, removable teeth
- 3 year manufacturers warranty
- Includes 3B smart anatomy
BodyTextAccessories: For information about 3B smart anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought