ContactUsLinkComponent
3B Scientific® Functional Physiological Skeleton
3B Scientific® Functional Physiological Skeleton
# de catalogue 470120-368
Fournisseur:  American 3B Scientific
MODEL PHYSIOLOGICAL SKELETON FUNCTIO
3B Scientific® Functional Physiological Skeleton
# de catalogue 470120-368
Fournisseur:  American 3B Scientific
Spécifications
  • Description:
    Functional Physiological Skeleton
  • Cat. no.:
    470120-368
  • Supplier No.:
    A15/3S
Spécifications
A propos de cet article
Show Your Students How The Skeleton Moves
  • Half The Size Of A Normal Skeleton
  • Available With Or Without Painted Muscle Locations
  • 3 Year Manufacturers Warranty
  • Includes 3B Smart Anatomy
This skeleton features an impressive new assembly of all the joints that enable it to make individual movements that are almost true-to-life. Demonstrate the complex movement sequences of the upper limb. The shoulder blades twist when the arms are raised. Even the position of spoke bones and ulna in relation to each other during internal and external rotation of the forearm can be shown clearly. The flexible spine of this skeleton can make natural movements (lateral bend, movement and stretching, rotation). When the knee joint is stretched and bent, thanks to the flexible assembly, the position of the knee cap can be demonstrated. The hand bones have been assembled on wire to show their natural position. The flexible assembly of the foot bones makes them maneuverable. Even the low level of maneuverability of the sacroiliac joint and the sacrococcygeal symphysis can be demonstrated. Now available on a stable metal hanging stand with 5 casters.
BodyTextAccessories: For information about 3B Smart Anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought