Lifting Demonstration Figure
# de catalogue 470118-960
Fournisseur: American 3B Scientific
Spécifications
- Description:Lifting Demonstration Figure
- Cat. no.:470118-960
- Supplier No.:W19007
Spécifications
A propos de cet article
A Great Way To Make Students More Aware Of The Effects Lifting Has On Their Spine.
- Flexible joints demonstrate proper and improper lifting
- Reinforcing key on base
- Large enough to be seen, but small enough to transport
Provide your students with a graphic demonstration of the effects of correct and incorrect lifting techniques on the spinal column. The figure lifts a weight from one of two positions. If the correct technique is used the spine stays in proper alignment. Using an incorrect technique causes the vertebrae of the spine to become distorted. Comes on base with illustrations of right and wrong lifting on the spine.
Customers Who Bought This Also Bought