Flow Control Adapters with Spherical Joints, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
These flow control adapters are for use with Chemglass absorption chromatography columns.
- Chem-Cap® metering plug
- Replacement plugs available
Adapters have a Chem-Cap® metering plug. Replacement plugs are available.
Customers Who Bought This Also Bought