Optimum Growth™ Inversion Transfer Caps, Thomson Instrument Company
Fournisseur: THOMSON INSTRUMENT CO
New Product
Certificats
A propos de cet article
The sterile Optimum Growth™ Transfer Caps and Inversion Transfer Caps allow for a simple aseptic transfer of media or cells to cell bags, bioreactors, and flasks of all makes.
These caps are designed for use with the Optimum Growth™ Flask product line. Simply replace the culture cap with the Transfer Cap and connect to the vessel of choice.
Customers Who Bought This Also Bought