Round German Glass Cover Slips, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Cover slips are manufactured from the highest quality hydrolytic Class 1 German borosilicate glass
All cover slips are #1 thickness. They have excellent resistance to chemicals and are ideal for neural tissue culture and hard-to-attach cells. These cover slips are exceptional for florescence and confocal microscopy. Sold in 1 oz. packages.
Customers Who Bought This Also Bought