Tacky® Roll Mops Hand Held 6-301, Liberty
# de catalogue 21924-076
Fournisseur: Liberty Industries
Small Business Enterprise
Spécifications
- Description:Tacky® Roll mop, Hand-Held, Model 6-301
- Cat. no.:21924-076
- Handle Length:9"
- Weight:12 oz.
- Supplier No.:10-1131
Spécifications
A propos de cet article
The Hand Held Tacky® Roll Mop has a 9" handle with one refill installed on the handle and is used on work surfaces, table tops, and hard-to-reach areas
It removes dirt, dust, debris, and lint from a variety of materials including glass, plastic, metal, cloth, and similar problem surfaces. The top sheet peels off to expose a fresh, clean surface when needed. The Hand Held Tacky Roll Mop has perforations about every 14" for easy access to a fresh sheet. For refills simply pull off the old core and replace it with a new roll.
Customers Who Bought This Also Bought